Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dihydroxy-2-(4-methoxyphenyl)acetophenone 98.0+%, TCI America™
SDP

Catalog No. D48161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48161G 1 g
D4816200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48161G Supplier TCI America Supplier No. D48161G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 487-49-0
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD00020119
InChI Key XHBZOAYMBBUURD-UHFFFAOYSA-N
Synonym ononetin, 1-2,4-dihydroxyphenyl-2-4-methoxyphenyl ethanone, 2',4'-dihydroxy-2-4-methoxyphenyl acetophenone, acetophenone, 2',4'-dihydroxy-2-p-methoxyphenyl, 2,4-dihydroxy-4'-methoxydeoxybenzoin, 1-2,4-dihydroxyphenyl-2-4-methoxyphenyl-ethanone, 1-2,4-dihydroxyphenyl-2-4-methoxyphenyl ethan-1-one, spectrum_000796, spectrum2_000808, spectrum3_001777
PubChem CID 259632
IUPAC Name 1-(2,4-dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)CC(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 258.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dihydroxy-2-(4-methoxyphenyl)acetophenone
RTECSNumber KM5775350
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.