Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxy-5-methoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. D45995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45995G 5 g
D45991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45995G Supplier TCI America Supplier No. D45995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6623-81-0
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00187903
InChI Key KELXHQACBIUYSE-UHFFFAOYSA-N
Synonym 5-Methoxy-2,4-pyrimidinediol
PubChem CID 81100
IUPAC Name 5-methoxy-1H-pyrimidine-2,4-dione
SMILES COC1=CNC(=O)NC1=O

Specifications

Melting Point 344°C
Color White
Quantity 5 g
Formula Weight 142.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxy-5-methoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.