Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzonitrile 99.0+%, TCI America™
SDP

Catalog No. D485425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D485425G 25 g
D48545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D485425G Supplier TCI America Supplier No. D485425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4107-65-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00001786
InChI Key RYRZSQQELLQCMZ-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dimethoxy, 2,4-dimethoxy-benzonitrile, 2,4-dimethoxybenzenecarbonitrile, pubchem13105, acmc-1anqc, 2.4-dimethoxybenzonitril, ksc497m9f, 2,4-dimethoxybenzonitrile
PubChem CID 77750
IUPAC Name 2,4-dimethoxybenzonitrile
SMILES COC1=CC(=C(C=C1)C#N)OC

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 163.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.