Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxyphenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. D494325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D494325G 25 g
D49435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D494325G Supplier TCI America Supplier No. D494325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33904-03-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00041058
InChI Key CNXSCEPTSXPBTP-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
PubChem CID 2736202
IUPAC Name 1-isothiocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=S)OC

Specifications

Melting Point 54°C
Color Yellow
Boiling Point 167°C
Quantity 25 g
UN Number 1759
Formula Weight 195.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxyphenyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.