Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzenethiol 96.0+%, TCI America™
SDP

Catalog No. D264825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D264825G 25 g
D26485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D264825G Supplier TCI America Supplier No. D264825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13616-82-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010019
InChI Key AMNLXDDJGGTIPL-UHFFFAOYSA-N
Synonym 2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiol
PubChem CID 83617
IUPAC Name 2,4-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)S)C

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 g
UN Number 1993
Formula Weight 138.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzenethiol
EINECSNumber (3)-1094
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.