Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylpentane 99.0+%, TCI America™
SDP

Catalog No. D076925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076925ML 25 mL
D07695ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D076925ML Supplier TCI America Supplier No. D076925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-08-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00008945
InChI Key BZHMBWZPUJHVEE-UHFFFAOYSA-N
Synonym pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane
PubChem CID 7907
IUPAC Name 2,4-dimethylpentane
SMILES CC(C)CC(C)C

Specifications

Color Colorless
Boiling Point 80°C
Quantity 25 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylpentane
EINECSNumber (2)-0007
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.