Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylstyrene, 97%, stabilized

Catalog No. AC271151000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271151000 100 g Glass bottle
AC271150050 5 g Glass bottle
AC271150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271151000 Supplier Thermo Scientific Chemicals Supplier No. 271151000

missing translation for 'validMainframeMsg'

CAS: 2234-20-0 | C10H12 | 132.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2234-20-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00014937
InChI Key OEVVKKAVYQFQNV-UHFFFAOYSA-N
Synonym 2,4-dimethylstyrene, 4-vinyl-m-xylene, styrene, 2,4-dimethyl, 1,3-dimethyl-4-vinylbenzene, 1-vinyl-2,4-dimethylbenzene, 1,3-dimethyl-4-ethenylbenzene, benzene, 1-ethenyl-2,4-dimethyl, unii-543es1o25e, styrene,4-dimethyl, acmc-1cb1s
PubChem CID 16694
IUPAC Name 1-ethenyl-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C=C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -64.0°C
Density 0.9060g/mL
Boiling Point 73.0°C to 74.0°C (10.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5410 to 1.5440
Linear Formula (CH3)2C6H3CH=CH2
Quantity 100 g
Specific Gravity 0.906
Stabilizer 0.05 % (ca.) t-butylcatechol
Formula Weight 132.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylstyrene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr
EINECSNumber 218-781-3
RTECSNumber WL4450000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.