Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitro-α-naphthol-7-sulfonic Acid, Na Salt, MP Biomedicals

Catalog No. ICN206436 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206436 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206436 Supplier MP Biomedicals, Inc Supplier No. 0520643601

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 846-70-8
Molecular Formula C10H4N2Na2O8S
Molecular Weight (g/mol) 358.188
InChI Key CTIQLGJVGNGFEW-UHFFFAOYSA-L
Synonym naphthol yellow s, acid yellow 1, c.i. acid yellow 1, sodium flavianate, naphthol yellow, acid yellow s, ext. FD&C yellow no. 7, unii-08f8s9o3i5, c.i. food yellow 1, ext. d & c yellow no. 7
PubChem CID 2724063
ChEBI CHEBI:87219
IUPAC Name disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
SMILES C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Formula Weight 358.19
Chemical Name or Material 2, 4-Dinitro-Alpha-Naphthol-7-Sulfonic Acid, Na Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.