accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 846-70-8

IUPAC Name: sodium 2,4-dinitro-7-sulfonatonaphthalen-1-olate
Synonyms: acid yellow 1
Molecular Weight (g/mol): 335.2
Molecular Formula: C10H4N2NaO8S
InChi Key: CGXSFOUXEQBDCJ-UHFFFAOYSA-L
SMILES: [Na+].[O-]C1=C(C=C(C2=CC=C(C=C12)S([O-])(=O)=O)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)