Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrofluorobenzene 99.0+%, TCI America™
SDP

Catalog No. A551225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A551225G 25 g
A55125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A551225G Supplier TCI America Supplier No. A551225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70-34-8
Molecular Formula C6H3FN2O4
Molecular Weight (g/mol) 186.098
MDL Number MFCD00007056
InChI Key LOTKRQAVGJMPNV-UHFFFAOYSA-N
Synonym 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
PubChem CID 6264
ChEBI CHEBI:53049
IUPAC Name 1-fluoro-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

Specifications

Melting Point 26°C
Boiling Point 296°C
Quantity 25 g
UN Number 2811
Formula Weight 186.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dinitrofluorobenzene [for HPLC Labeling]
EINECSNumber (3)-0456
RTECSNumber CZ7800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.