Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenol Solution, 1000μg/mL in methanol, Ultra Scientific

Catalog No. USPH1401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PH-1401 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PH-1401 Supplier Agilent Technologies Supplier No. PH1401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-28-5 , 67-56-1
Molecular Formula C6H4N2O5
Molecular Weight (g/mol) 184.107
InChI Key UFBJCMHMOXMLKC-UHFFFAOYSA-N
PubChem CID 1493
ChEBI CHEBI:42017
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])O

Specifications

CAS Max % 99.87, 0.13
Analyte or Component Names Methanol,2,4-Dinitrophenol
Element Methanol,2,4-Dinitrophenol
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material 2, 4-Dinitrophenol Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.