Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2,4-Dioxo-3,4-dihydro-2H-pyrimidin-1-yl)-acetic acid

Catalog No. CDS012581 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS012581 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS012581 Supplier Sigma Aldrich Supplier No. CDS01258150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-4-Methylcyclohexanol is an active Aedes triseriatus oviposition attractant.trans-4-Methylcyclohexanol was used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.trans-4-Methylcyclohexanol is the major metabolite formed by biological regioselective reduction of 4-methylcyclohexanone by Glomerella cingulata.

Specifications

Molecular Formula C6H6N2O4
Linear Formula C6H6N2O4
MDL Number MFCD00458007
Quantity 10 g
Molecular Weight (g/mol) 170.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.