Learn More
2,4-Diphenyl-1,3-thiazole-5-sulfonyl chloride, ≥97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| IUPAC Name | 2,4-diphenyl-1,3-thiazole-5-sulfonyl chloride |
| Formula Weight | 335.83 |
| Percent Purity | 97% |
| Chemical Name or Material | 2, 4-Diphenyl-1, 3-thiazole-5-sulfonyl chloride |
Safety and Handling
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage. Contact with water liberates toxic gas. Reacts violently with water. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.