Learn More
Sigma Organic Chemistry 2- 4-Fluorophenoxy etha | 1G | MFCD11841289
Supplier: Sigma Organic Chemistry CBR005981G

2- 4-Fluorophenoxy etha , 1G
About this Item:
MDL #: MFCD11841289
Molecular Weight: 251.28
UNSPSC Code: 12352200
Chemical Formula: C9H14FNO4S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.