Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Fluorophenyl)thiophene 95.0+%, TCI America™
SDP

Catalog No. F07241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07241G 1 g
F07245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07241G Supplier TCI America Supplier No. F07241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58861-48-6
Molecular Formula C10H7FS
Molecular Weight (g/mol) 178.224
MDL Number MFCD06802535
InChI Key PURJRGMZIKXDMW-UHFFFAOYSA-N
PubChem CID 21894783
IUPAC Name 2-(4-fluorophenyl)thiophene
SMILES C1=CSC(=C1)C2=CC=C(C=C2)F

Specifications

Melting Point 53°C
Color Yellow
Quantity 1 g
Formula Weight 178.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.