Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Mesitylenedisulfonyl Dichloride 98.0+%, TCI America™
SDP

Catalog No. M118625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M118625G 25 g
M11865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M118625G Supplier TCI America Supplier No. M118625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68985-08-0
Molecular Formula C9H10Cl2O4S2
Molecular Weight (g/mol) 317.20
MDL Number MFCD00009796
InChI Key LNXKRGBQKLXFTB-UHFFFAOYSA-N
Synonym 2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride
PubChem CID 3620081
IUPAC Name 2,4,6-trimethylbenzene-1,3-disulfonyl dichloride
SMILES CC1=CC(C)=C(C(C)=C1S(Cl)(=O)=O)S(Cl)(=O)=O

Specifications

Melting Point 125°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 317.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Mesitylenedisulfonyl Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.