Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxyphenyl)pyridine 97.0+%, TCI America™
SDP

Catalog No. M21011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21011G 1 g
M21015G 5 g
M210125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M21011G Supplier TCI America Supplier No. M21011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5957-90-4
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00092333
InChI Key QUMXRZNAUFKBAS-UHFFFAOYSA-N
Synonym 4-(2-Pyridyl)anisole
PubChem CID 220239
IUPAC Name 2-(4-methoxyphenyl)pyridine
SMILES COC1=CC=C(C=C1)C2=CC=CC=N2

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 185.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methoxyphenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.