Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Acetate 98.0+%, TCI America™
SDP

Catalog No. M184825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M184825G 25 g
M18485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M184825G Supplier TCI America Supplier No. M184825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 656-53-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.24
MDL Number MFCD00005338
InChI Key CRTCWNPLKVVXIX-UHFFFAOYSA-N
Synonym 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole
PubChem CID 61192
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
SMILES CC(=O)OCCC1=C(C)N=CS1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 g
Formula Weight 185.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Acetate
EINECSNumber (5)-6975
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.