Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methyl-5-thiazolyl)ethyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. M248925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M248925G 25 g
M24895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M248925G Supplier TCI America Supplier No. M248925G

Chemical Identifiers

CAS 94159-31-6
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.30
MDL Number MFCD09032913
InChI Key GDRZNYCKSKHESZ-UHFFFAOYSA-N
Synonym Butyric Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Butyryloxyethyl)-4-methylthiazole
PubChem CID 3023838
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl butanoate
SMILES CCCC(=O)OCCC1=C(C)N=CS1

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 213.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl Butyrate
EINECSNumber (5)-3412
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.