Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methylphenyl)ethanol, 99%

Catalog No. AAB2091806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2091806 5 g
AAB2091803 1 g
AAB2091814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2091806 Supplier Thermo Scientific Chemicals Supplier No. B2091806
Add to Cart

CAS: 699-02-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002904
InChI Key DAVFJRVIVZOKKS-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol
PubChem CID 95294
IUPAC Name 2-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)CCO

Specifications

Density 0.98
Boiling Point 244°C to 245°C
Flash Point 107°C (225°F)
Refractive Index 1.526
Quantity 5 g
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2-(4-Methylphenyl)ethanol
EINECSNumber 211-824-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.