Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-(4-Methylpiperazin-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC36009CB
Change view
Click to view available options
Quantity:
1 mg
250 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC36009CB 250 mg
CC36009DA 1 mg
CC36009EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC36009CB Supplier Thermo Scientific Chemicals Supplier No. CC36009CB
Add to Cart

Chemical Identifiers

CAS 123987-12-2
Molecular Formula C12H19N2O
Molecular Weight (g/mol) 207.30
MDL Number MFCD03407383
InChI Key TWPYBKBPHCMUIS-UHFFFAOYSA-O
Synonym 2-4-methylpiperazin-1-yl phenyl methanol, 2-4-methylpiperazin-1-yl benzyl alcohol, 2-4-methylpiperazino benzyl alcohol, 2-4-methylpiperazin-1yl phenyl methanol, pubchem12999, 2-4-methylpiperazin benzyl alcohol, 2-4-n-methylpiperazinyl benzyl alcohol, 2-4-methyl-1-piperazinyl phenyl methanol, 2-4-methylpiperazinyl phenyl methan-1-ol
PubChem CID 2795567
SMILES C[NH+]1CCN(CC1)C1=CC=CC=C1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 4-[2-(hydroxymethyl)phenyl]-1-methylpiperazin-1-ium
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material [2-(4-Methylpiperazin-1-yl)phenyl]methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.