Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Nitrophenyl)ethyl Bromide 98.0+%, TCI America™
SDP

Catalog No. N059110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059110G 10 g
N059125G 25 g
N0591100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N059110G Supplier TCI America Supplier No. N059110G

Chemical Identifiers

CAS 5339-26-4
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00007386
InChI Key NTURQZFFJDCTMZ-UHFFFAOYSA-N
Synonym 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
PubChem CID 79266
IUPAC Name 1-(2-bromoethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCBr)[N+](=O)[O-]

Specifications

Melting Point 69°C
Quantity 10 g
Formula Weight 230.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenyl)ethyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.