Learn More
Sigma Organic Chemistry 2- 4-Nitrophenyl thiazo | 1G | MFCD12114473
Supplier: Sigma Organic Chemistry BOG001881G

2- 4-Nitrophenyl thiazo , 1G
About this Item:
MDL #: MFCD12114473
Molecular Weight: 264.26
UNSPSC Code: 12352200
Chemical Formula: C11H8N2O4S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.