Learn More
Sigma Organic Chemistry 2- 4-Piperidinyl propan | 1G | MFCD18483419
Supplier: Sigma Organic Chemistry CBR014091G

2- 4-Piperidinyl propan , 1G
About this Item:
MDL #: MFCD18483419
Molecular Weight: 193.67
UNSPSC Code: 12352200
Chemical Formula: C8H16ClNO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.