Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Piperidyl)-2-propanol 98.0+%, TCI America™
SDP

Catalog No. P147025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P147025G 25 g
P14701G 1 g
P14705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P147025G Supplier TCI America Supplier No. P147025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22990-34-7
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD06797159
InChI Key IIXYTWTZMGUQPT-UHFFFAOYSA-N
Synonym 2-(4-Piperidyl)isopropyl Alcohol
PubChem CID 5200321
IUPAC Name 2-(piperidin-4-yl)propan-2-ol
SMILES CC(C)(O)C1CCNCC1

Specifications

Melting Point 136°C
Color White
Quantity 25 g
Formula Weight 143.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Piperidyl)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.