Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic Acid 98.0+%, TCI America™
SDP

Catalog No. B2322100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2322100MG 100 mg
B23221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2322100MG Supplier TCI America Supplier No. B2322100MG

Chemical Identifiers

CAS 308815-83-0
Molecular Formula C15H23NO6Si
Molecular Weight (g/mol) 341.44
MDL Number MFCD04117911
InChI Key UXUZQGZIXUXYHB-UHFFFAOYSA-N

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 341.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[4-(tert-Butyldimethylsilyloxymethyl)-2-nitrophenoxy]acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.