Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene 98.0+%, TCI America™
SDP

Catalog No. B155425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B155425G 25 g
B15545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B155425G Supplier TCI America Supplier No. B155425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7128-64-5
Molecular Formula C26H26N2O2S
Molecular Weight (g/mol) 430.566
MDL Number MFCD00005774
InChI Key AIXZBGVLNVRQSS-UHFFFAOYSA-N
Synonym bbot, 2,5-bis 5-tert-butyl-2-benzoxazolyl thiophene, uvitex ob, 2,5-bis 5-tert-butyl benzo d oxazol-2-yl thiophene, bbot 150, 2,5-bis 5-tert-butylbenzoxazol-2-yl thiophene, benzoxazole, 2,2'-2,5-thiophenediyl bis 5-1,1-dimethylethyl, unii-w7ur9129gp, fluorescent brightener 184, 2,2'-2,5-thiophenediyl bis 5-tert-butylbenzoxazole
PubChem CID 292429
IUPAC Name 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
SMILES CC(C)(C)C1=CC2=C(C=C1)OC(=N2)C3=CC=C(S3)C4=NC5=C(O4)C=CC(=C5)C(C)(C)C

Specifications

Melting Point 201°C
Quantity 25 g
Formula Weight 430.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
EINECSNumber (5)-3934
RTECSNumber DM4888332
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.