Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™
SDP

Catalog No. B38681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38681G 1 g
B38685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38681G Supplier TCI America Supplier No. B38681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 927384-46-1
Molecular Formula C14H10BBrN2S
Molecular Weight (g/mol) 329.022
InChI Key POCAQBXWVHDOHU-UHFFFAOYSA-N
Synonym 2-Bromo-5-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)thiophene, 2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
PubChem CID 56973702
SMILES B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC=C(S4)Br

Specifications

Melting Point 141°C
Color Yellow
Quantity 1 g
Formula Weight 329.02
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.