Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di-tert-amylhydroquinone 93.0+%, TCI America™
SDP

Catalog No. D029925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D029925G 25 g
D0299500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D029925G Supplier TCI America Supplier No. D029925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79-74-3
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00027029
InChI Key CZNRFEXEPBITDS-UHFFFAOYSA-N
Synonym 2,5-Di-tert-pentylhydroquinone
PubChem CID 6610
IUPAC Name 2,5-bis(2-methylbutan-2-yl)benzene-1,4-diol
SMILES CCC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)CC

Specifications

Melting Point 179°C
Quantity 25 g
Formula Weight 250.38
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di-tert-amylhydroquinone
EINECSNumber (3)-0553
RTECSNumber MX6300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.