Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3,4-ethylenedioxythiophene 98.0+%, TCI America™
SDP

Catalog No. D40565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40565G 5 g
D40561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40565G Supplier TCI America Supplier No. D40565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 174508-31-7
Molecular Formula C6H4Br2O2S
Molecular Weight (g/mol) 299.96
MDL Number MFCD01651770
InChI Key FHMRWRBNAIDRAP-UHFFFAOYSA-N
Synonym 5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine
PubChem CID 3834570
IUPAC Name 5,7-dibromo-2H,3H-thieno[3,4-b][1,4]dioxine
SMILES BrC1=C2OCCOC2=C(Br)S1

Specifications

Melting Point 96°C
Quantity 5 g
Formula Weight 299.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3,4-ethylenedioxythiophene
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.