Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-fluoropyridine, 95%

Catalog No. AAH6110803
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6110803 1 g
AAH61108MD 250 mg
AAH6110806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6110803 Supplier Thermo Scientific Chemicals Supplier No. H6110803

CAS: 156772-60-0 | C5H2Br2FN | 254.884 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156772-60-0
Molecular Formula C5H2Br2FN
Molecular Weight (g/mol) 254.884
MDL Number MFCD06659497
InChI Key QEENLQKOTDHQEW-UHFFFAOYSA-N
PubChem CID 22412832
IUPAC Name 2,5-dibromo-3-fluoropyridine
SMILES C1=C(C(=NC=C1Br)Br)F

Specifications

Color White
Quantity 1 g
UN Number UN2811
Synonym 2,5-dibromo-3-fluoro-pyridine, pyridine, 2,5-dibromo-3-fluoro, pubchem17547, acmc-209deo, ksc529c7p, pyridine,2,5-dibromo-3-fluoro, abbypharma ap-30-7594, 2,5-bis bromanyl-3-fluoranyl-pyridine, 2-bromo-3-fluoro-5-bromopyridine
Formula Weight 254.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,5-Dibromo-3-fluoropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.