Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-tetradecylthiophene 97.0+%, TCI America™
SDP

Catalog No. D52411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52411G 1 g
D52415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52411G Supplier TCI America Supplier No. D52411G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 160096-74-2
Molecular Formula C18H30Br2S
Molecular Weight (g/mol) 438.31
MDL Number MFCD30742898
InChI Key IYQPIBPAABQINF-UHFFFAOYSA-N
PubChem CID 101678000
IUPAC Name 2,5-dibromo-3-tetradecylthiophene
SMILES CCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 438.31
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dibromo-3-tetradecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.