Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide 98.0+%, TCI America™
SDP

Catalog No. D50211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50211G 1 g
D5021200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50211G Supplier TCI America Supplier No. D50211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1231160-83-0
Molecular Formula C14H17Br2NO2S
Molecular Weight (g/mol) 423.163
MDL Number MFCD23703120
InChI Key AOZLCBYWDXFKCJ-UHFFFAOYSA-N
Synonym 1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione
PubChem CID 58261078
IUPAC Name 1,3-dibromo-5-(2-ethylhexyl)thieno[3,4-c]pyrrole-4,6-dione
SMILES CCCCC(CC)CN1C(=O)C2=C(SC(=C2C1=O)Br)Br

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
Formula Weight 423.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.