Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. D343925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D343925G 25 g
D34395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D343925G Supplier TCI America Supplier No. D343925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 610-71-9
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016494
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
PubChem CID 11891
IUPAC Name 2,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 279.92
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DG6290000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.