Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. D476925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D476925G 25 g
D47695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D476925G Supplier TCI America Supplier No. D476925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7657-09-2
Molecular Formula C7H3Br2F3
Molecular Weight (g/mol) 303.90
MDL Number MFCD00013554
InChI Key VWKFJAOCLPPQGR-UHFFFAOYSA-N
Synonym 1,4-dibromo-2-trifluoromethyl benzene, 2,5-dibromobenzotrifluoride, benzene, 1,4-dibromo-2-trifluoromethyl, 2,5-dibromo trifluoromethyl benzene, 1,4-dibromo-2-trifluoromethyl-benzene, toluene, 2,5-dibromo-alpha,alpha,alpha-trifluoro, pubchem9537, 2,5-dibromobenzotrifloride, 2,5-dibromo-, ,-trifluorotoluene
PubChem CID 82108
IUPAC Name 1,4-dibromo-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 207°C
Quantity 25 g
Formula Weight 303.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dibromobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.