Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromopyridine 99.0+%, TCI America™
SDP

Catalog No. D1217500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1217500G 500 g
D12175G 5 g
D121725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1217500G Supplier TCI America Supplier No. D1217500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 624-28-2
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00006221
InChI Key ZHXUWDPHUQHFOV-UHFFFAOYSA-N
Synonym 2,5-dibromo pyridine, pyridine, 2,5-dibromo, 2,5-dibrompyridin, 2,5-dibromo-pyridine, 2,5-dibromopyridin, 3,6-dibromopyridine, 2,5,-dibromopyridine, 2,5-dibrompyridine, 2,5 dibromopyridine, pyridine,5-dibromo
PubChem CID 69353
IUPAC Name 2,5-dibromopyridine
SMILES BrC1=CC=C(Br)N=C1

Specifications

Melting Point 93°C
Color White-Yellow
Boiling Point 235°C
Quantity 500 g
Formula Weight 236.89
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.