Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetoacetanilide, 98%, Thermo Scientific™

Catalog No. AAA1670014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1670014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1670014 Supplier Thermo Scientific Chemicals Supplier No. A1670014

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-72-6
Molecular Formula C10H9Cl2NO2
Molecular Weight (g/mol) 246.087
MDL Number MFCD00018520
InChI Key HLMZZYYGOKOOTA-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetoacetanilide, n-2,5-dichlorophenyl-3-oxobutanamide, acetoacetic acid-2,5-dichloroanilide, butanamide, n-2,5-dichlorophenyl-3-oxo, acetoacetanilide, 2',5'-dichloro, acmc-1cg2h, 2,5-dichloroacetoacetanilide, acetoacetanilide,5'-dichloro, n-acetoacetyl-2,5-dichloroaniline, butanamide,5-dichlorophenyl-3-oxo
PubChem CID 74890
IUPAC Name N-(2,5-dichlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 94°C to 96°C
Quantity 25 g
Formula Weight 246.09
Percent Purity 98%
Chemical Name or Material 2',5'-Dichloroacetoacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-060-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.