Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetophenone 98.0+%, TCI America™
SDP

Catalog No. D250225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D250225G 25 g
D25025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D250225G Supplier TCI America Supplier No. D250225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 128°C
Quantity 25 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,5′-Dichloroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.