Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorofluorobenzene 98.0+%, TCI America™
SDP

Catalog No. D251325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D251325G 25 g
D25135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D251325G Supplier TCI America Supplier No. D251325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 348-59-4
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.99
MDL Number MFCD00060656
InChI Key BJQTYCQGIXZSNM-UHFFFAOYSA-N
Synonym 2,5-dichlorofluorobenzene, benzene, 1,4-dichloro-2-fluoro, 1,4-dichloro-2-fluoro-benzene, 2,5-dichloro-1-fluorobenzene, 1,4-dichlor-2-fluorbenzol, 2,5-dichloro-fluorobenzene, pubchem3454, acmc-1ae1b, 2,5-dichloro fluorobenzene, 1-fluor-2,5-dichlorbenzol
PubChem CID 136173
IUPAC Name 1,4-dichloro-2-fluorobenzene
SMILES FC1=CC(Cl)=CC=C1Cl

Specifications

Boiling Point 168°C
Quantity 25 g
Formula Weight 164.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dichlorofluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.