Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloroisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D2599500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2599500MG 500 mg
D25995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2599500MG Supplier TCI America Supplier No. D2599500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88912-26-9
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00466640
InChI Key GFOVTTQVBDEYPP-UHFFFAOYSA-N
Synonym 2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
PubChem CID 2736087
IUPAC Name 2,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)Cl)C(=O)O

Specifications

Color White-Yellow
Quantity 500 mg
Formula Weight 192.00
Percent Purity ≥98.0% (T,GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.