Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloropyridine 99.0+%, TCI America™
SDP

Catalog No. D160725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D160725G 25 g
D16075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D160725G Supplier TCI America Supplier No. D160725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

Melting Point 61°C
Color White
Boiling Point 98°C
Quantity 25 g
Formula Weight 147.99
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloropyridine
RTECSNumber US8225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.