Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. D404525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D404525G 25 g
D40455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D404525G Supplier TCI America Supplier No. D404525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22536-67-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00673131
InChI Key CEJAHXLRNZJPQH-UHFFFAOYSA-N
Synonym 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci
PubChem CID 590641
IUPAC Name 2,5-dichloropyrimidine
SMILES C1=C(C=NC(=N1)Cl)Cl

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.