Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorosulfanilic Acid 98.0+%, TCI America™
SDP

Catalog No. D041525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041525G 25 g
D0415500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D041525G Supplier TCI America Supplier No. D041525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-50-6
Molecular Formula C6H5Cl2NO3S
Molecular Weight (g/mol) 242.07
MDL Number MFCD00025159
InChI Key SJCTXIKOXTUQHC-UHFFFAOYSA-N
Synonym 2,5-Dichloroaniline-4-sulfonic Acid
PubChem CID 66619
IUPAC Name 4-amino-2,5-dichlorobenzenesulfonic acid
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)O)Cl)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 242.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorosulfanilic Acid
EINECSNumber (3)-2085
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.