Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorotoluene 98.0+%, TCI America™
SDP

Catalog No. D215725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D215725G 25 g
D21575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D215725G Supplier TCI America Supplier No. D215725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19398-61-9
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000609
InChI Key KFAKZJUYBOYVKA-UHFFFAOYSA-N
Synonym 2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
PubChem CID 29572
IUPAC Name 1,4-dichloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 g
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dichlorotoluene
EINECSNumber (3)-0078
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.