Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester, 96%

Catalog No. AAH5902503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5902503 1 g
AAH59025MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5902503 Supplier Thermo Scientific Chemicals Supplier No. H5902503
Add to Cart

CAS: 303006-90-8 | C18H26B2F2O4 | 366.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303006-90-8
Molecular Formula C18H26B2F2O4
Molecular Weight (g/mol) 366.019
MDL Number MFCD12407211
InChI Key PJCSUAIUPWCQRS-UHFFFAOYSA-N
Synonym 2,2'-2,5-difluoro-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,5-difluoro-1,4-phenylenediboronic acid, pinacol ester, 2-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,5-difluoro-1,4-phenylenediboronic acid,pinacol ester, 2,2'-2,5-difluoro-p-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,2-2,5-difluoro-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 53216814
IUPAC Name 2-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)B3OC(C(O3)(C)C)(C)C)F

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Formula Weight 366.02
Percent Purity 96%
Chemical Name or Material 2,5-Difluorobenzene-1,4-diboronic acid bis(pinacol) ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.