Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzophenone 96.0+%, TCI America™
SDP

Catalog No. D35985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35985G Supplier TCI America Supplier No. D35985G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85068-36-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00009899
InChI Key HSCUAAMDKDZZKG-UHFFFAOYSA-N
Synonym 2,5-difluorobenzophenone, 2,5-difluorophenyl phenyl methanone, 2,5-difluoro-phenyl-phenyl-methanone, methanone, 2,5-difluorophenyl phenyl, 2,5-difluoro diphenylmethanone, 2,5-difluorophenyl-phenylmethanone, 2,5-difluorophenyl phenyl ketone, pubchem4214, 2.5-difluorobenzophenone, 2,5-difluoro benzophenone
PubChem CID 522826
IUPAC Name (2,5-difluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)F)F

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 218.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.