Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D343625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D343625G 25 g
D34365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D343625G Supplier TCI America Supplier No. D343625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 193353-34-3
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD01863171
InChI Key KTOJGSDLJNUAEP-UHFFFAOYSA-N
Synonym 2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
PubChem CID 2734335
IUPAC Name (2,5-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)F)F)(O)O

Specifications

Quantity 25 g
Formula Weight 157.91
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.