Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diformylfuran 98.0+%, TCI America™
SDP

Catalog No. D2408100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2408100MG 100 mg
D24081G 1 g
D24085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2408100MG Supplier TCI America Supplier No. D2408100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 823-82-5
Molecular Formula C6H4O3
Molecular Weight (g/mol) 124.10
MDL Number MFCD00671517
InChI Key PXJJKVNIMAZHCB-UHFFFAOYSA-N
Synonym 2,5-Furandicarboxaldehyde
PubChem CID 69980
ChEBI CHEBI:83385
IUPAC Name furan-2,5-dicarbaldehyde
SMILES O=CC1=CC=C(O1)C=O

Specifications

Melting Point 110°C
Quantity 100 mg
Formula Weight 124.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diformylfuran
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.