Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydrofuran 98.0+%, TCI America™
SDP

Catalog No. D0551100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0551100ML 100 mL
D055125ML 25 mL
D0551500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0551100ML Supplier TCI America Supplier No. D0551100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1708-29-8
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00003219
InChI Key ARGCQEVBJHPOGB-UHFFFAOYSA-N
Synonym furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
PubChem CID 15570
IUPAC Name 2,5-dihydrofuran
SMILES C1C=CCO1

Specifications

Color Yellow
Boiling Point 67°C
Quantity 100 mL
UN Number 1993
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dihydrofuran
EINECSNumber (9)-0731
RTECSNumber LU0660000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.