Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dihydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. D195525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D195525G 25 g
D19555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D195525G Supplier TCI America Supplier No. D195525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 490-78-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002343
InChI Key WLDWSGZHNBANIO-UHFFFAOYSA-N
Synonym 2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy
PubChem CID 10279
IUPAC Name 1-(2,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 207°C
Color Yellow
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,5′-Dihydroxyacetophenone
EINECSNumber (3)-1250
RTECSNumber AM7700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.